missing translation for 'onlineSavingsMsg'
Learn More

1-Pyrrolidinecarbonyl chloride, 97%

Product Code. 10538332 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10538332 25 g Glass Bottle 25g
10639574 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10538332 Supplier Thermo Scientific Acros Supplier No. 418650250
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1192-63-8 | C5H8ClNO | 133.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1192-63-8
Molecular Formula C5H8ClNO
Molecular Weight (g/mol) 133.58
MDL Number MFCD00051321
InChI Key XACWJIQLDLUFSR-UHFFFAOYSA-N
Synonym 1-pyrrolidinecarbonyl chloride, pyrrolidinecarbonyl chloride, 1-pyrrolidinecarbonylchloride, pyrrolidine-1-carbonylchloride, pyrrolidinocarbonyl chloride, 1pyrrolidinecarbonyl chloride, pyrrolidine carbonyl chloride, acmc-1bo54, 1-pyrrolidine carbonylchloride, 1-pyrolidine carbonyl chloride
PubChem CID 70937
IUPAC Name pyrrolidine-1-carbonyl chloride
SMILES C1CCN(C1)C(=O)Cl

Specifications

Boiling Point 75.0°C (1.0 mmHg)
CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4918 to 1.4938
Density 1.2090g/mL
Quantity 25 g
Flash Point 98°C
Formula Weight 133.58
Percent Purity 97%
Specific Gravity 1.209
Physical Form Liquid
Chemical Name or Material 1-Pyrrolidinecarbonyl chloride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Pyrrolidinecarbonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.