missing translation for 'onlineSavingsMsg'
Learn More

1-Pyrrolidinecarbodithioic Acid Ammonium Salt, TRC

Product Code. 30426278
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30426278 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30426278 Supplier Toronto Research Chemicals Supplier No. TRCP997985100G
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Pyrrolidinecarbodithioic Acid Ammonium Salt
CAS 5108-96-3
Molecular Formula C5H12N2S2
Quantity 100 g
Synonym 1-Pyrrolidinecarbodithioic acid','ammonium salt (8CI,9CI), 1-Pyrrolidinedithiocarboxylic acid ammonium salt, APDC, Ammonium 1-pyrrolidinecarbodithioate, Ammonium 1-pyrrolidinedithiocarboxylate, Ammonium N,N-tetramethylenedithiocarbamate, Ammonium pyrrolidine-1-dithiocarbamate, Ammonium pyrrolidine-N-dithiocarbamate, Ammonium pyrrolidinecarbodithioate, Ammonium pyrrolidinedithiocarboxylate, Pyrrolidine-N-dithiocarbamic Acid Ammonium
InChI Formula InChI=1S/C5H9NS2.H3N/c7-5(8)6-3-1-2-4-6;/h1-4H2,(H,7,8);1H3
SMILES [NH4+].[S-]C(=S)N1CCCC1
IUPAC Name azanium;pyrrolidine-1-carbodithioate
Molecular Weight (g/mol) 164.29
Formula Weight 164.0442
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.