missing translation for 'onlineSavingsMsg'
Learn More

1-Pyrenecarboxaldehyde, 98+%

Product Code. 11353016
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11353016 5 g 5g
11363016 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11353016 Supplier Thermo Scientific Alfa Aesar Supplier No. L10835.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3029-19-4 | C17H10O | 230.266 g/mol

1-Pyrenecarboxaldehyde is a important intermediate used in agrochemical, pharmaceutical and dyestuff field. It is used as a highly polarity-sensitive fluorophore for SNP typing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pyrenecarboxaldehyde is a important intermediate used in agrochemical, pharmaceutical and dyestuff field. It is used as a highly polarity-sensitive fluorophore for SNP typing.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3029-19-4
Molecular Formula C17H10O
Molecular Weight (g/mol) 230.266
MDL Number MFCD00004139
InChI Key RCYFOPUXRMOLQM-UHFFFAOYSA-N
Synonym 1-pyrenecarboxaldehyde, 1-formylpyrene, 1-pyrenealdehyde, pyrenecarboxaldehyde, 3-formylpyrene, 3-pyrenealdehyde, 3-pyrenylaldehyde, pyrene-1-aldehyde, 3-pyrenecarboxaldehyde, 1-pyrenecarbaldehyde
PubChem CID 232848
IUPAC Name pyrene-1-carbaldehyde
SMILES C1=CC2=C3C(=C1)C=CC4=C(C=CC(=C43)C=C2)C=O

Specifications

Melting Point 125°C to 128°C
Quantity 5 g
Beilstein 1874889
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 230.27
Percent Purity 99%
Chemical Name or Material 1-Pyrenecarboxaldehyde
compliance-icons
Product Identifier
  • 1-Pyrenecarboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Pyrene-1-carbaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.