Learn More
1-Pyrenebutyric acid, 97%
Description
1-Pyrenebutyric acid is a fluorescent probe used to study proteins, lipids, nucleic acids and other biological systems. It acts as a UV excitable fluorophore that has concentration-dependent fluorescence. It is also employed as a probe for bovine serum. 1-pyrene butyric acid modified nitrogen-doped carbon nanotubes (PBA/NCNTs) nanocomposites can be used as a biosensor to detect glucose in human serum sample.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Pyrenebutyric acid is a fluorescent probe used to study proteins, lipids, nucleic acids and other biological systems. It acts as a UV excitable fluorophore that has concentration-dependent fluorescence. It is also employed as a probe for bovine serum. 1-pyrene butyric acid modified nitrogen-doped carbon nanotubes (PBA/NCNTs) nanocomposites can be used as a biosensor to detect glucose in human serum sample.
Solubility
Partially soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 184°C to 186°C |
| Quantity | 1 g |
| Beilstein | 2140554 |
| Solubility Information | Partially soluble in water. |
| IUPAC Name | 4-(pyren-1-yl)butanoic acid |
| Formula Weight | 288.35 |
| Percent Purity | 97% |
| Chemical Name or Material | 1-Pyrenebutyric acid |
Safety and Handling
- 1-Pyrenebutyric acid
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contain: 1-Pyrenebutanoic acid
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.