missing translation for 'onlineSavingsMsg'
Learn More

1-Pyrenebutyric acid, 97%

Product Code. p-7023825
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11325129

Brand: Thermo Scientific Alfa Aesar A17760.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3443-45-6 | C20H16O2 | 288.35 g/mol

1-Pyrenebutyric acid is a fluorescent probe used to study proteins, lipids, nucleic acids and other biological systems. It acts as a UV excitable fluorophore that has concentration-dependent fluorescence. It is also employed as a probe for bovine serum. 1-pyrene butyric acid modified nitrogen-doped carbon nanotubes (PBA/NCNTs) nanocomposites can be used as a biosensor to detect glucose in human serum sample.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pyrenebutyric acid is a fluorescent probe used to study proteins, lipids, nucleic acids and other biological systems. It acts as a UV excitable fluorophore that has concentration-dependent fluorescence. It is also employed as a probe for bovine serum. 1-pyrene butyric acid modified nitrogen-doped carbon nanotubes (PBA/NCNTs) nanocomposites can be used as a biosensor to detect glucose in human serum sample.

Solubility
Partially soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3443-45-6
Molecular Formula C20H16O2
Molecular Weight (g/mol) 288.35
MDL Number MFCD00004141
InChI Key QXYRRCOJHNZVDJ-UHFFFAOYSA-N
Synonym 1-pyrenebutyric acid, 4-pyren-1-yl butanoic acid, 1-pyrenebutanoic acid, 1-pyrenylbutyric acid, pyrene-3-butyric acid, pyrenebutanoic acid, pyrene-1-butyric acid, pyrenebutyric acid, 4-1-pyrenyl butyric acid, gamma-3-pyrenyl butyric acid
PubChem CID 76977
SMILES OC(=O)CCCC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34

Specifications

Melting Point 184°C to 186°C
Quantity 1 g
Beilstein 2140554
Solubility Information Partially soluble in water.
IUPAC Name 4-(pyren-1-yl)butanoic acid
Formula Weight 288.35
Percent Purity 97%
Chemical Name or Material 1-Pyrenebutyric acid
compliance-icons
Product Identifier
  • 1-Pyrenebutyric acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 1-Pyrenebutanoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.