missing translation for 'onlineSavingsMsg'
Learn More

1-Propynyltri-n-butyltin, 96%

Product Code. 11369036
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11369036 1 g 1g
11379036 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11369036 Supplier Thermo Scientific Alfa Aesar Supplier No. L20201.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 64099-82-7 | C15H30Sn | 329.115 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64099-82-7
Molecular Formula C15H30Sn
Molecular Weight (g/mol) 329.115
MDL Number MFCD01863650
InChI Key KCQJLTOSSVXOCC-UHFFFAOYSA-N
Synonym tributyl 1-propynyl tin, tributyl prop-1-yn-1-yl stannane, tributyl prop-1-ynyl stannane, tributylpropynylstannane, 1-propynyltri-n-butyltin, stannane, tributyl-1-propynyl, tributyl-1-propyn-1-ylstannane, tributyl-prop-1-ynyl-stannane, propynyltributyltin;, ch3ccsnbu3
PubChem CID 2733165
IUPAC Name tributyl(prop-1-ynyl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C#CC

Specifications

Density 1.082
Boiling Point 277°C
Flash Point >110°C (230°F)
Linear Formula H3CC≡CSn[(CH2)3CH3]3
Refractive Index 1.483
Quantity 1 g
UN Number UN2788
Formula Weight 329.12
Percent Purity 96%
Chemical Name or Material 1-Propynyltri-n-butyltin
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Propynyltri-n-butyltin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Tributylpropynylstannane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.