missing translation for 'onlineSavingsMsg'
Learn More

1-Propylpiperazine Dihydrobromide, TRC

Product Code. 30412069
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30412069 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30412069 Supplier Toronto Research Chemicals Supplier No. TRCP838518500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Propylpiperazine Dihydrobromide
CAS 64262-23-3
Molecular Formula C7H16N2 . 2HBr
Quantity 500 mg
Synonym Piperazine Impurity 17
InChI Formula InChI=1S/C7H16N2.2BrH/c1-2-5-9-6-3-8-4-7-9;;/h8H,2-7H2,1H3;2*1H
SMILES Br.Br.CCCN1CCNCC1
IUPAC Name 1-propylpiperazine;dihydrobromide
Molecular Weight (g/mol) 290.03
Formula Weight 287.9837
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.