missing translation for 'onlineSavingsMsg'
Learn More

1-Propylboronic acid, 98%, Thermo Scientific Chemicals

£51.30 - £692.00

Chemical Identifiers

CAS 17745-45-8
Molecular Formula C3H9BO2
Molecular Weight (g/mol) 87.91
MDL Number MFCD01074564
InChI Key JAQOMSTTXPGKTN-UHFFFAOYSA-N
Synonym n-propylboronic acid, 1-propaneboronic acid, propaneboronic acid, boronic acid, propyl, 1-propylboronic acid, 1-propaneboronic, n-propylboronc acd, propyl boronic acid, pubchem6019
PubChem CID 351065
IUPAC Name propylboronic acid
SMILES CCCB(O)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11364346
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L19965.03
1 g
£51.30
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11374346
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L19965.06
5 g
£175.00
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11384346
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L19965.14
25 g
£692.00
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

17745-45-8
87.91
JAQOMSTTXPGKTN-UHFFFAOYSA-N
351065
CCCB(O)O
C3H9BO2
MFCD01074564
n-propylboronic acid, 1-propaneboronic acid, propaneboronic acid, boronic acid, propyl, 1-propylboronic acid, 1-propaneboronic, n-propylboronc acd, propyl boronic acid, pubchem6019
propylboronic acid

Specifications

17745-45-8
C3H9BO2
1731884
n-propylboronic acid, 1-propaneboronic acid, propaneboronic acid, boronic acid, propyl, 1-propylboronic acid, 1-propaneboronic, n-propylboronc acd, propyl boronic acid, pubchem6019
CCCB(O)O
87.91
87.91
1-Propylboronic acid
105°C to 107°C
MFCD01074564
Air sensitive; Hygroscopic
JAQOMSTTXPGKTN-UHFFFAOYSA-N
propylboronic acid
351065
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Keep cold; Store under Argon

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only