missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Propylamine, 99+%, Thermo Scientific Chemicals
56.86 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 mL
1 L
Description
1-Propylamine is used in chemical analysis and to make other chemicals. It is used as finishing agent of drugs, paints, rubber, fiber, textile and resin. Also it can be used as petroleum additive and preservative. It is used in the production of fiber and engineering plastics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -83°C |
Color | Colorless |
Density | 0.719 g/mL |
pH | 12.6 |
Boiling Point | 48°C to 49°C |
Flash Point | -12°C (10°F) |
Assay Percent Range | >99+% |
Odor | Amine-like |
Refractive Index | 1.389 |
Quantity | 1 L |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'