missing translation for 'onlineSavingsMsg'
Learn More

1-Propylamine, 98%, Thermo Scientific Chemicals

Catalog Number 11434693
Click to view available options
Quantity:
500 mL
2500 mL
This item is not returnable. View return policy
107-10-8
C3H9N
59.112
MFCD00008205
WGYKZJWCGVVSQN-UHFFFAOYSA-N
propylamine, 1-propanamine, 1-propylamine, propanamine, n-propylamine, 1-aminopropane, monopropylamine, mono-n-propylamine, propyl amine, n-propyl amine
7852
CHEBI:39870
propan-1-amine
CCCN
This item is not returnable. View return policy
107-10-8
C3H9N
59.112
MFCD00008205
WGYKZJWCGVVSQN-UHFFFAOYSA-N
propylamine, 1-propanamine, 1-propylamine, propanamine, n-propylamine, 1-aminopropane, monopropylamine, mono-n-propylamine, propyl amine, n-propyl amine
7852
CHEBI:39870
propan-1-amine
CCCN

1-Propylamine is used in chemical analysis and to make other chemicals, used as a solvent in organic synthesis. And it is used as finishing agent of drugs, paints, rubber, fiber, textile and resin. Additionally, it can be used as petroleum additive and preservative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Propylamine is used in chemical analysis and to make other chemicals, used as a solvent in organic synthesis. And it is used as finishing agent of drugs, paints, rubber, fiber, textile and resin. Additionally, it can be used as petroleum additive and preservative.

Solubility
Miscible with water, alcohol, ether

Notes
Miscible with water, alcohol, ether
TRUSTED_SUSTAINABILITY
Melting Point -83°C
Density 0.719
pH 12.6
Boiling Point 48°C to 49°C
Flash Point −12°C (10°F)
Odor Amine-like
Refractive Index 1.389
Quantity 500 mL
UN Number UN1277
Beilstein 1098243
Sensitivity Air Sensitive
Merck Index 14,7843
Solubility Information Miscible with water,alcohol,ether
Formula Weight 59.11
Percent Purity 98%
Chemical Name or Material 1-Propylamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'