missing translation for 'onlineSavingsMsg'
Learn More

(1-Propyl)triphenylphosphonium bromide, 99%

Product Code. 11409833
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409833 50 g 50g
11419833 250 g 250g
11499823 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11409833 Supplier Thermo Scientific Alfa Aesar Supplier No. A12968.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6228-47-3 | C21H22BrP | 385.29 g/mol

It is employed as a catalyst for the carboxylation of styrene oxide and as a reactant involved in epoxidation of arylalkenes, boration of allylic epoxides, Wittig reactions, selective dimerization of α-olefins, and synthesis of indoles. It has also been shown to be part of Phosphonium salts, having antiviral activity against influenza virus A.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a catalyst for the carboxylation of styrene oxide and as a reactant involved in epoxidation of arylalkenes, boration of allylic epoxides, Wittig reactions, selective dimerization of α-olefins, and synthesis of indoles. It has also been shown to be part of Phosphonium salts, having antiviral activity against influenza virus A.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents, humidity and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6228-47-3
Molecular Formula C21H22BrP
Molecular Weight (g/mol) 385.29
MDL Number MFCD00011843
InChI Key XMQSELBBYSAURN-UHFFFAOYSA-M
Synonym propyltriphenylphosphonium bromide, triphenyl propyl phosphonium bromide, 1-propyl triphenylphosphonium bromide, n-propyltriphenylphosphonium bromide, triphenyl propyl phosphanium bromide, n-propyl triphenylphosphonium bromide, propyl triphenyl phosphonium chloride, acmc-1bff2, propyltriphenylphosphoniumbromide
PubChem CID 80374
IUPAC Name triphenyl(propyl)phosphanium;bromide
SMILES [Br-].CCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 236°C to 240°C
Quantity 50 g
Beilstein 3599726
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 385.29
Percent Purity 99%
Chemical Name or Material (1-Propyl)triphenylphosphonium bromide
compliance-icons
Product Identifier
  • (1-Propyl)triphenylphosphonium bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Triphenylpropylphosphonium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.