missing translation for 'onlineSavingsMsg'
Learn More

1-Propanol, puriss. p.a., Solstice

Product Code. 15683980
Change view
Click to view available options
Quantity:
1 L
2.5 L
160 kg
Unit Size:
1 L
160kg
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15683980 160 kg 160kg
15693980 1 L 1 L
15603990 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15683980 Supplier Honeywell Riedel-de Haën Supplier No. 33538160KG
Request Bulk or Custom Formats

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 71-23-8
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00002941
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
Synonym 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
PubChem CID 1031
ChEBI CHEBI:28831
IUPAC Name propan-1-ol
SMILES CCCO

Specifications

Density 0.804g/mL at 25°C
Quantity 160 kg
Explosion Limit 0.137
Flash Point 22°C (71.6°F)
Ignition Point 700°F
Linear Formula CH3CH2CH2OH
Vapor Pressure 14.9 mmHg (20°C), 10 mmHg (147°C)
UN Number UN1274
Beilstein 1098242
Formula Weight 60.1g/mol
Percent Purity ≥99.5% (GC)
Grade puriss. p.a.
Chemical Name or Material 1-Propanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.