Learn More
1-Propanol, 99+%, Thermo Scientific Chemicals
£29.10 - £47.20
Chemical Identifiers
CAS | 71-23-8 |
---|---|
Molecular Formula | C3H8O |
Molecular Weight (g/mol) | 60.10 |
MDL Number | MFCD00002941 |
InChI Key | BDERNNFJNOPAEC-UHFFFAOYSA-N |
Synonym | 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol |
PubChem CID | 1031 |
ChEBI | CHEBI:28831 |
IUPAC Name | propan-1-ol |
SMILES | CCCO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11498727
|
Thermo Scientific Alfa Aesar
A19902.AP |
500 mL |
£29.10
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11408737
|
Thermo Scientific Alfa Aesar
A19902.0F |
2500 mL |
£47.20
2500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Propanol is used as a solvent and an entrainer in azeotropic distillation. It is used as a precursor to prepare propyl acetate, propylamide and propyl halides by reacting with acetic acid, ammonia and halogens respectively. It is actively used as a solvent in inks for flexographic printing, which is a technique used for printing. It is added to cleaners, floor polishes and metal degreasing fluids and is used as an additional solvent in the manufacture of adhesives. It enhances the drying characteristics of alkyd resins, electrodeposition paints and baking finishes in coating industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Propanol is used as a solvent and an entrainer in azeotropic distillation. It is used as a precursor to prepare propyl acetate, propylamide and propyl halides by reacting with acetic acid, ammonia and halogens respectively. It is actively used as a solvent in inks for flexographic printing, which is a technique used for printing. It is added to cleaners, floor polishes and metal degreasing fluids and is used as an additional solvent in the manufacture of adhesives. It enhances the drying characteristics of alkyd resins, electrodeposition paints and baking finishes in coating industry.
Solubility
Miscible with water, alcohols, ketones, aldehydes, ethers, glycols, aromatic and aliphatic hydrocarbons.
Notes
Highly flammable. Incompatible with alkali metals, alkaline earths, aluminum, oxidizing agents, nitro compounds.
Chemical Identifiers
71-23-8 | |
60.10 | |
BDERNNFJNOPAEC-UHFFFAOYSA-N | |
1031 | |
propan-1-ol |
C3H8O | |
MFCD00002941 | |
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol | |
CHEBI:28831 | |
CCCO |
Specifications
-127°C | |
0.804 | |
15°C (59°F) | |
1.385 | |
MFCD00002941 | |
1098242 | |
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol | |
BDERNNFJNOPAEC-UHFFFAOYSA-N | |
propan-1-ol | |
1031 | |
60.1 | |
1-Propanol |
71-23-8 | |
97°C | |
Alcohol-like | |
C3H8O | |
UN1274 | |
14,7842 | |
Miscible with water,alcohols,ketones,aldehydes,ethers,glycols,aromatic and aliphatic hydrocarbons. | |
CCCO | |
60.10 | |
CHEBI:28831 | |
≥99% |
Safety and Handling
GHS H Statement
H225-H318-H336
Highly flammable liquid and vapour.
Causes serious eye damage.
May cause drowsiness or dizziness.
P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P370+P378q-P501c
H225-H318-H336
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: N-PROPANOL
missing translation for 'einecsNumber' : 200-746-9
missing translation for 'rtecsNumber' : UH8225000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only