missing translation for 'onlineSavingsMsg'
Learn More

1-Propanethiol, 98%

Product Code. 11440580
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440580 500 mL 500mL
11385707 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11440580 Supplier Thermo Scientific Alfa Aesar Supplier No. 043420.AP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 107-03-9 | C3H8S | 76.157 g/mol

1-Propanethiol, is used widely in research areas. This compound has a wide range of applications including cell biology, materials science, nanoscience, self-assembled monolayers (SAMs), nanoelectronics, and contact printing. It is also used as a chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-03-9
Molecular Formula C3H8S
Molecular Weight (g/mol) 76.157
MDL Number MFCD00004900
InChI Key SUVIGLJNEAMWEG-UHFFFAOYSA-N
Synonym 1-propanethiol, propanethiol, n-propylmercaptan, propyl mercaptan, n-propyl mercaptan, n-propylthiol, 1-propylmercaptan, propylthiol, 1-mercaptopropane, 1-propyl mercaptan
PubChem CID 7848
ChEBI CHEBI:8473
IUPAC Name propane-1-thiol
SMILES CCCS

Specifications

Melting Point -113°C
Color Colorless
Density 0.8410 g/mL
Boiling Point 67°C to 68°C
Flash Point −20°C (4°F)
Odor Stench
Linear Formula CH3CH2CH2SH
Refractive Index 1.438
Quantity 500 mL
UN Number UN2402
Sensitivity Air Sensitive
Solubility Information It is soluble in water (1.9g/L) at 25°C. It is also soluble in alcohol,acetone,benzene,ether,hydrocarbons.
Formula Weight 76.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Propanethiol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.