missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Propanephosphonic acid cyclic anhydride, 50 wt.% sol. in ethyl acetate, Thermo Scientific Chemicals
86.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
50mL; Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS | 68957-94-8, 141-78-6 |
Color | Brown to Yellow |
Density | 1.0690g/mL |
Boiling Point | 77.2°C |
Flash Point | −5°C |
Packaging | Glass bottle |
Molecular Formula | C9H21O6P3 |
Refractive Index | 1.4500 to 1.4540 (20°C, 589nm) |
MDL Number | MFCD00006583 |
Quantity | 50 mL |
Show More |
Safety and Handling
Product Identifier
- 1-Propanephosphonic acid cyclic anhydride, 50 wt.% solution in ethyl acetate
- Danger
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'