missing translation for 'onlineSavingsMsg'
Learn More

1-Propan-1,1,2,2,3,3,3-d7-ol, TRC

Product Code. 30421183
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30421183 250 mg -
30417704 50 mg -
30411949 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30421183 Supplier Toronto Research Chemicals Supplier No. TRCP757403250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102910-31-6
Molecular Formula C3 2H7 H O
Molecular Weight (g/mol) 67.14
Synonym 1-Propanol D7, 1,1,2,2,3,3,3-Heptadeuteriopropan-1-ol, 1-Propan-1,1,2,2,3,3,3-d7-ol (9CI)
IUPAC Name 1,1,2,2,3,3,3-heptadeuteriopropan-1-ol
SMILES [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C3H8O/c1-2-3-4/h4H,2-3H2,1H3/i1D3,2D2,3D2
Formula Weight 67.1015
Chemical Name or Material 1-Propanol D7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.