missing translation for 'onlineSavingsMsg'
Learn More

1-Piperonylpiperazine, 97%, ACROS Organics™

Brand:  Acros Organics 208931000

Code : 29

Additional Details : CAS Number : 32231-06-4 Weight : 0.15000kg Transport : UN number : 3259 Chem class : 1b Pack group : III


Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10114550

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

1-Piperonylpiperazine
32231-06-4
100.0
97%
220.27
97%
96.0
≥96.0%
C12H16N2O2
MFCD00005834

Specifications

1-Piperonylpiperazine
32231-06-4
100.0
≥96.0%
Authentic
220.27
Glass bottle
38° to 42°C
97%
96.0
Crystalline Solid
97%
C12H16N2O2
MFCD00005834
White to yellow or yellow-brownish
100g
Videos
SDS