missing translation for 'onlineSavingsMsg'
Learn More

1-Piperazinepropionitrile

Product Code. 11410270
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410270 1 g 1g
11420270 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11410270 Supplier Thermo Scientific Alfa Aesar Supplier No. H50710.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 34064-86-3 | C7H13N3 | 139.202 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34064-86-3
Molecular Formula C7H13N3
Molecular Weight (g/mol) 139.202
MDL Number MFCD02093566
InChI Key MVOFPBMQTXKONX-UHFFFAOYSA-N
Synonym 3-piperazin-1-yl propanenitrile, 3-piperazin-1-yl-propionitrile, 1-piperazinepropionitrile, 3-1-piperazinyl-propionitril, 1-2-cyanoethyl piperazine, 3-piperazin-1-yl propionitrile, 3-1-piperazino propionitrile, 3-1-piperazinyl propionitrile, 3-1-piperazinyl-propionitrile, 3-piperazinylpropanenitrile
PubChem CID 2052050
IUPAC Name 3-piperazin-1-ylpropanenitrile
SMILES C1CN(CCN1)CCC#N

Specifications

Quantity 1 g
UN Number UN3276
Formula Weight 139.2
Chemical Name or Material 1-Piperazinepropionitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.