missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Piperazine-d8-acetic Acid 2HCl, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | 2-(Piperazin-1-yl)acetic Acid-D8 (piperazine-D8) Dihydrochloride |
| Molecular Formula | C6 D8 H4 N2 O2 . 2 Cl H |
| Quantity | 0.05 g |
| Synonym | 1-Piperazine-d8-acetic acid hydrochloride (1:2), 2-(Piperazin-1-yl-2,2,3,3,5,5,6,6-d8)acetic acid hydrochloride (1:2), 2-(Piperazin-1-yl)acetic acid-d8 dihydrochloride, 2-(Piperazin-1-yl)acetic acid-D8 (piperazine-D8) dihydrochloride |
| InChI Formula | InChI=1 S/C6H12N2O2.2 ClH/c9-6(10)5-8-3-1-7-2-4-8;;/h7H,1-5H2,(H,9,10);2*1 H/i1D2,2D2,3D2,4D2;; |
| SMILES | Cl.Cl.[2 H]C1([2 H])NC([2 H])([2 H])C([2 H])([2 H])N(CC(=O)O)C1([2 H])[2 H] |
| IUPAC Name | 2-(2,2,3,3,5,5,6,6-octadeuteriopiperazin-1-yl)acetic acid;dihydrochloride |
| Molecular Weight (g/mol) | 225.1429 |
| Formula Weight | 224.0934 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction