missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylvinylboronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Product Code. 15499967
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15499967 1 g 1g
15409977 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15499967 Supplier Thermo Scientific Alfa Aesar Supplier No. H53216.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143825-84-7
Molecular Formula C14H19BO2
Molecular Weight (g/mol) 230.114
MDL Number MFCD06659923
InChI Key RMGBWPMWUZSIMH-UHFFFAOYSA-N
Synonym 4,4,5,5-tetramethyl-2-1-phenylvinyl-1,3,2-dioxaborolane, 1-phenylvinylboronic acid pinacol ester, 2-1-phenylethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-1-phenylethenyl-1,3,2-dioxaborolane, 1-phenylvinylboronic acid, pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-1-phenylethenyl, 1-phenylvinylboronic acid pinacolester, 2-1-phenylvinyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 2-1-phenylvinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 11031671
IUPAC Name 4,4,5,5-tetramethyl-2-(1-phenylethenyl)-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C(=C)C2=CC=CC=C2

Specifications

Melting Point 40°C to 42°C
Quantity 1 g
Formula Weight 230.11
Percent Purity 95%
Chemical Name or Material 1-Phenylvinylboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.