missing translation for 'onlineSavingsMsg'
Learn More

1-(Phenylsulfonyl)-1H-indole-2-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10691473
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10691473 1 g 1g
10794953 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10691473 Supplier Thermo Scientific Maybridge Supplier No. CC03304DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 80360-23-2
Molecular Formula C15H11NO3S
Molecular Weight (g/mol) 285.32
MDL Number MFCD03086091
InChI Key NWVOBVXBWUGTTO-UHFFFAOYSA-N
Synonym 1-phenylsulfonyl-1h-indole-2-carbaldehyde, 1-benzenesulfonyl indole-2-carbaldehyde, 1-benzenesulfonyl-1h-indole-2-carbaldehyde, 2-formyl-1-phenylsulfonyl-1h-indole, 2-formyl-1h-indol-1-yl phenyl sulfone, 1-phenylsulphonyl-1h-indole-2-carboxaldehyde, 1h-indole-2-carboxaldehyde,1-phenylsulfonyl, phenylsulfonyl-1h-indole-2-carbaldehyde
PubChem CID 2776223
IUPAC Name 1-(benzenesulfonyl)indole-2-carbaldehyde
SMILES O=CC1=CC2=CC=CC=C2N1S(=O)(=O)C1=CC=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 285.32
Percent Purity 97%
Chemical Name or Material 1-(Phenylsulfonyl)-1H-indole-2-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.