missing translation for 'onlineSavingsMsg'
Learn More

1-(Phenylsulfonyl)-1H-indole-2-carbaldehyde, 97%, Thermo Scientific™

Catalog Number p-2694532
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
80360-23-2
C15H11NO3S
285.32
MFCD03086091
NWVOBVXBWUGTTO-UHFFFAOYSA-N
1-phenylsulfonyl-1h-indole-2-carbaldehyde, 1-benzenesulfonyl indole-2-carbaldehyde, 1-benzenesulfonyl-1h-indole-2-carbaldehyde, 2-formyl-1-phenylsulfonyl-1h-indole, 2-formyl-1h-indol-1-yl phenyl sulfone, 1-phenylsulphonyl-1h-indole-2-carboxaldehyde, 1h-indole-2-carboxaldehyde,1-phenylsulfonyl, phenylsulfonyl-1h-indole-2-carbaldehyde
2776223
1-(benzenesulfonyl)indole-2-carbaldehyde
O=CC1=CC2=CC=CC=C2N1S(=O)(=O)C1=CC=CC=C1
This item is not returnable. View return policy
80360-23-2
C15H11NO3S
285.32
MFCD03086091
NWVOBVXBWUGTTO-UHFFFAOYSA-N
1-phenylsulfonyl-1h-indole-2-carbaldehyde, 1-benzenesulfonyl indole-2-carbaldehyde, 1-benzenesulfonyl-1h-indole-2-carbaldehyde, 2-formyl-1-phenylsulfonyl-1h-indole, 2-formyl-1h-indol-1-yl phenyl sulfone, 1-phenylsulphonyl-1h-indole-2-carboxaldehyde, 1h-indole-2-carboxaldehyde,1-phenylsulfonyl, phenylsulfonyl-1h-indole-2-carbaldehyde
2776223
1-(benzenesulfonyl)indole-2-carbaldehyde
O=CC1=CC2=CC=CC=C2N1S(=O)(=O)C1=CC=CC=C1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 285.32
Percent Purity 97%
Chemical Name or Material 1-(Phenylsulfonyl)-1H-indole-2-carbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'