missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylsemicarbazide, 99%

Product Code. 11406046
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11406046 25 g 25g
11416046 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11406046 Supplier Thermo Scientific Alfa Aesar Supplier No. B22331.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103-03-7 | C7H9N3O | 151.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-03-7
Molecular Formula C7H9N3O
Molecular Weight (g/mol) 151.17
MDL Number MFCD00007942
InChI Key AVKHCKXGKPAGEI-UHFFFAOYSA-N
Synonym phenicarbazide, 1-phenylsemicarbazide, 2-phenylhydrazinecarboxamide, phenicarbazid, phenylsemicarbazide, carbaphen, kryogenin, phenygenine, febrimin, 1-carbamyl-2-phenylhydrazine
PubChem CID 61002
ChEBI CHEBI:82521
IUPAC Name anilinourea
SMILES NC(=O)NNC1=CC=CC=C1

Specifications

Melting Point 171°C to 173°C
Odor Odorless
Quantity 25 g
Formula Weight 151.17
Percent Purity 99%
Chemical Name or Material 1-Phenylsemicarbazide
compliance-icons compliance-icons
Product Identifier
  • 1-Phenylsemicarbazide
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Phenicarbazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.