missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Phenylpyrrolidine, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
1-Phenylpyrrolidine are used as flooding intestinal coccidiosis, flooding Xiao NET intermediate.It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Phenylpyrrolidine are used as flooding intestinal coccidiosis, flooding Xiao NET intermediate.It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Not miscible in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents. Store cold 2-8°C.
1-Phenylpyrrolidine are used as flooding intestinal coccidiosis, flooding Xiao NET intermediate.It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Not miscible in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents. Store cold 2-8°C.
Specifications
Boiling Point | 133°C to 134°C (19 mmHg) |
Refractive Index | 1.582 |
Quantity | 25 g |
Beilstein | 117030 |
Solubility Information | Not miscible in water. |
Formula Weight | 147.22 |
Percent Purity | ≥98% |
Chemical Name or Material | 1-Phenylpyrrolidine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'