missing translation for 'onlineSavingsMsg'
Learn More

1-(Phenylmethyl)-4-piperidinamine, TRC

Product Code. 30412371
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30412371 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30412371 Supplier Toronto Research Chemicals Supplier No. TRCP33584050G
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(Phenylmethyl)-4-piperidinamine
CAS 50541-93-0
Molecular Formula C12H18N2
Quantity 50 g
Synonym (1-Benzylpiperidin-4-yl)amine, 1-(Phenylmethyl)-4-piperidinamine, 1-Benzyl-4-aminopiperidine, 1-Benzyl-4-piperidinamine, 1-Benzyl-4-piperidinylamine, 1-Benzylpiperidine-4-amine, 2-Amino-1-benzylpiperidine, 4-Amino-1-(phenylmethyl)piperidine, 4-Amino-1-benzylpiperidine, 4-Amino-N-benzylpiperidine, N-Benzyl-4-aminopiperidine, N-Benzyl-4-piperidinamine
InChI Formula InChI=1S/C12H18N2/c13-12-6-8-14(9-7-12)10-11-4-2-1-3-5-11/h1-5,12H,6-10,13H2
SMILES NC1CCN(Cc2ccccc2)CC1
IUPAC Name 1-benzylpiperidin-4-amine
Molecular Weight (g/mol) 190.28
Formula Weight 190.147
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.