missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylisatin, 98%

Product Code. 11377746
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11377746 5 g 5g
11367746 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377746 Supplier Thermo Scientific Alfa Aesar Supplier No. L11388.06
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 723-89-7 | C14H9NO2 | 223.231 g/mol

1-Phenylisatin is a reactant for preparation of indolin-2,3-dione Schiff bases as potential antimycobacterial agents, antipyretic agents, antiproliferative agents, phenylacetamides as anticonvulsant agents and selective Galanin GAL3 receptor antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Phenylisatin is a reactant for preparation of indolin-2,3-dione Schiff bases as potential antimycobacterial agents, antipyretic agents, antiproliferative agents, phenylacetamides as anticonvulsant agents and selective Galanin GAL3 receptor antagonists.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 723-89-7
Molecular Formula C14H9NO2
Molecular Weight (g/mol) 223.231
MDL Number MFCD00082681
InChI Key UWCPWBIMRYXUOU-UHFFFAOYSA-N
Synonym 1-phenylisatin, 1-phenylindoline-2,3-dione, 1-phenyl-1h-indole-2,3-dione, 1h-indole-2,3-dione, 1-phenyl, 1-phenyl-indole-2,3-dione, indole-2,3-dione, 1-phenyl, 1h-indole-2,3-dione, 1-phenyl-9ci, 1-phenyl-2,3-dihydro-1h-indole-2,3-dione, phenylisatin, n-phenylisatin
PubChem CID 12884
IUPAC Name 1-phenylindole-2,3-dione
SMILES C1=CC=C(C=C1)N2C3=CC=CC=C3C(=O)C2=O

Specifications

Melting Point 139°C to 142°C
Quantity 5 g
Beilstein 164531
Solubility Information Insoluble in water.
Formula Weight 223.23
Percent Purity 98%
Chemical Name or Material 1-Phenylisatin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.