missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-1-Phenylethanol, 97%

Product Code. 11461478
Change view
Click to view available options
Quantity:
10,000 g
2500 g
500 g
Unit Size:
10000g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11461478 10,000 g 10000g
11444934 500 g 500g
11451478 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11461478 Supplier Thermo Scientific Alfa Aesar Supplier No. A13837.0C
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 98-85-1 | C8H10O | 122.167 g/mol

The Arthrobacter sp. is able to grow with (+ I-,(-)-1-phenylethanol or the racemic mixture as sole source of carbon. Growth is most rapid with the (-)-isomer, doubling time 12 h. Lipases show good activity and, in some cases, improved enantioselectivity when employed in pure ionic liquids for dynamic kinetic resolution of 1-phenylethanol by transesterification.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The Arthrobacter sp. is able to grow with (+ I-,(-)-1-phenylethanol or the racemic mixture as sole source of carbon. Growth is most rapid with the (-)-isomer, doubling time 12 h. Lipases show good activity and, in some cases, improved enantioselectivity when employed in pure ionic liquids for dynamic kinetic resolution of 1-phenylethanol by transesterification.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-85-1
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.167
MDL Number MFCD00004508
InChI Key WAPNOHKVXSQRPX-UHFFFAOYSA-N
Synonym methylphenyl carbinol, 1-phenylethan-1-ol, alpha-methylbenzyl alcohol, styralyl alcohol, 1-phenethyl alcohol, styrallyl alcohol, methylphenylcarbinol, 1-phenylethyl alcohol, phenylmethylcarbinol, 1-hydroxyethyl benzene
PubChem CID 7409
ChEBI CHEBI:669
IUPAC Name 1-phenylethanol
SMILES CC(C1=CC=CC=C1)O

Specifications

Melting Point 19°C to 20°C
Boiling Point 203°C to 205°C
Quantity 10,000 g
UN Number UN2937
Formula Weight 122.17
Percent Purity 97%
Chemical Name or Material (+/-)-1-Phenylethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.