missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylcyclopropanemethylamine, 97%, Thermo Scientific™

Product Code. 10225854
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10225854 1 g Amber Glass Bottle 1g
10527484 250 mg Amber Glass Bottle 250mg
10158794 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10225854 Supplier Thermo Scientific Maybridge Supplier No. MO08400DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 935-42-2
Molecular Formula C10H13N
Molecular Weight (g/mol) 147.22
MDL Number MFCD06213079
InChI Key FUEMGCALFIHXAS-UHFFFAOYSA-N
Synonym 1-phenylcyclopropyl methanamine, 1-phenylcyclopropyl methylamine, 1-phenylcyclopropanemethylamine, 1-1-phenylcyclopropyl methanamine, cyclopropanemethanamine, 1-phenyl, phenylcyclopropyl methylamine, 1-phenylcyclopropylmethylamine, chembl61251, 1-phenylcyclopropyl methyl amine
PubChem CID 70287
IUPAC Name (1-phenylcyclopropyl)methanamine
SMILES NCC1(CC1)C1=CC=CC=C1

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 147.22
Percent Purity 97%
Chemical Name or Material 1-phenylcyclopropanemethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.