missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylcyclopentanecarboxylic acid, 98%

Product Code. 10159462 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10159462 5 g Glass bottle 5g
10226070 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10159462 Supplier Thermo Scientific Acros Supplier No. 161140050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 77-55-4 | C12H14O2 | 190.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-55-4
Molecular Formula C12H14O2
Molecular Weight (g/mol) 190.24
MDL Number MFCD00001372
InChI Key RHPCYZLXNNRRMB-UHFFFAOYSA-N
Synonym 1-phenylcyclopentanecarboxylic acid, 1-phenyl-1-cyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 1-phenyl, 1-phenyl-cyclopentanecarboxylic acid, 1-phenylcyclopentane carboxylic acid, cyclopentanecarboxylicacid, 1-phenyl, pubchem22923, acmc-209pao, phenylcyclopentanecarboxylic acid, phenylcyclopentane carboxylic acid
PubChem CID 66167
IUPAC Name 1-phenylcyclopentane-1-carboxylic acid
SMILES OC(=O)C1(CCCC1)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 157°C to 161°C
Color Brown-Gray to Cream
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H5C5H8CO2H
Quantity 5 g
Beilstein 09, II, 418
Formula Weight 190.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1-Phenylcyclopentanecarboxylic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Phenylcyclopentanecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.