missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylcyclobutanecarbonitrile, 95%, Thermo Scientific™

Product Code. 10635782 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10635782

Marke: Thermo Scientific Acros 154950050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Chemical Identifiers

CAS 14377-68-5
MDL Number MFCD00019258
InChI Key DHIDUDPFTZJPCQ-UHFFFAOYSA-N
Synonym 1-phenylcyclobutanecarbonitrile, cyclobutanecarbonitrile, 1-phenyl, 1-phenyl-cyclobutanecarbonitrile, acmc-1c523, cyclobutanecarbonitrile,1-phenyl, 1-phenyl-1-cyclobutanecarbonitrile, 1-phenyl-1-cyclobutane carbonitrile
PubChem CID 84400
IUPAC Name 1-phenylcyclobutane-1-carbonitrile
SMILES C1CC(C1)(C#N)C2=CC=CC=C2

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Yellow
Density 1.0300g/mL
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass Bottle
Molecular Formula C11H11N
Linear Formula C6H5C4H6CN
Refractive Index 1.5313 to 1.5333
Quantity 5 g
Molecular Weight (g/mol) 157.216
Formula Weight 157.21
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1-Phenylcyclobutanecarbonitrile, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Phenylcyclobutanecarbonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.