missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-3-pyrazolidinone 97%, ACROS Organics™

Product Code. 10617322 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10617322 25 g Plastic bottle 25g
10627512 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Produktkod. 10617322 Leverantör Thermo Scientific Acros Leverantörsnummer 155980250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

Kemiska identifierare

CAS 92-43-3
Molecular Formula C9H10N2O
Molecular Weight (g/mol) 162.192
MDL Number MFCD00003094
InChI Key CMCWWLVWPDLCRM-UHFFFAOYSA-N
Synonym phenidone, 1-phenyl-3-pyrazolidone, fenidon, 1-phenyl-3-pyrazolidinone, 3-pyrazolidinone, 1-phenyl, 1-phenyl-3-oxopyrazolidine, 2-pyrazolin-3-ol, 1-phenyl, pyrazolidinone, 1-phenyl, 1-phenyl-pyrazolidin-3-one, unii-h0u5612p6k
PubChem CID 7090
IUPAC Name 1-phenylpyrazolidin-3-one
SMILES C1CN(NC1=O)C2=CC=CC=C2

Specifikationer

CAS Min % 96.0
CAS Max % 100.0
Melting Point 119°C to 123°C
Color Beige to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Plastic bottle
Quantity 25 g
Beilstein 24, 2
Merck Index 15, 7419
Formula Weight 162.19
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material 1-Phenyl-3-pyrazolidinone, 97%
Visa mer Visa mindre
compliance-icons compliance-icons
Produktbeteckning
  • 1-Phenyl-3-pyrazolidinone
Signalord
  • Warning
Riskkategorier
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
Faroangivelser
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Skyddsangivelse
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P273-Avoid release to the environment.
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.