missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-3-pyrazolidinone, 97%, Thermo Scientific Chemicals

Catalog Number p-7081903
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
92-43-3
C9H10N2O
162.192
MFCD00003094
CMCWWLVWPDLCRM-UHFFFAOYSA-N
phenidone, 1-phenyl-3-pyrazolidone, fenidon, 1-phenyl-3-pyrazolidinone, 3-pyrazolidinone, 1-phenyl, 1-phenyl-3-oxopyrazolidine, 2-pyrazolin-3-ol, 1-phenyl, pyrazolidinone, 1-phenyl, 1-phenyl-pyrazolidin-3-one, unii-h0u5612p6k
7090
1-phenylpyrazolidin-3-one
C1CN(NC1=O)C2=CC=CC=C2
This item is not returnable. View return policy
92-43-3
C9H10N2O
162.192
MFCD00003094
CMCWWLVWPDLCRM-UHFFFAOYSA-N
phenidone, 1-phenyl-3-pyrazolidone, fenidon, 1-phenyl-3-pyrazolidinone, 3-pyrazolidinone, 1-phenyl, 1-phenyl-3-oxopyrazolidine, 2-pyrazolin-3-ol, 1-phenyl, pyrazolidinone, 1-phenyl, 1-phenyl-pyrazolidin-3-one, unii-h0u5612p6k
7090
1-phenylpyrazolidin-3-one
C1CN(NC1=O)C2=CC=CC=C2

Phenidone inhibits both the LO (lipoxygenase) and Cox (cyclooxygenase) pathways, the synthesis of Fos-related antigen protein, and is described as an anti-inflammatory and anti-oxidant compound. Phenidone has been shown to block neurotoxicity induced by kainate, and weakens oxidative stress induced by lipopolysaccharide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenidone inhibits both the LO (lipoxygenase) and Cox (cyclooxygenase) pathways, the synthesis of Fos-related antigen protein, and is described as an anti-inflammatory and anti-oxidant compound. Phenidone has been shown to block neurotoxicity induced by kainate, and weakens oxidative stress induced by lipopolysaccharide.

Solubility
Soluble in water (1:10 hot), methanol (0.1 mg/mL), DMSO (75 mg/mL), and ethanol.

Notes
Store in cool place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 120°C to 123°C
Quantity 50 g
UN Number UN2811
Beilstein 131856
Merck Index 14,7309
Formula Weight 162.19
Percent Purity 97%
Chemical Name or Material 1-Phenyl-3-pyrazolidinone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'