missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Phenyl-3-pyrazolidinone, 97%, Thermo Scientific Chemicals
17.53 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
50 g
250 g
1000 g
Description
Phenidone inhibits both the LO (lipoxygenase) and Cox (cyclooxygenase) pathways, the synthesis of Fos-related antigen protein, and is described as an anti-inflammatory and anti-oxidant compound. Phenidone has been shown to block neurotoxicity induced by kainate, and weakens oxidative stress induced by lipopolysaccharide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phenidone inhibits both the LO (lipoxygenase) and Cox (cyclooxygenase) pathways, the synthesis of Fos-related antigen protein, and is described as an anti-inflammatory and anti-oxidant compound. Phenidone has been shown to block neurotoxicity induced by kainate, and weakens oxidative stress induced by lipopolysaccharide.
Solubility
Soluble in water (1:10 hot), methanol (0.1 mg/mL), DMSO (75 mg/mL), and ethanol.
Notes
Store in cool place. Keep away from oxidizing agents.
Phenidone inhibits both the LO (lipoxygenase) and Cox (cyclooxygenase) pathways, the synthesis of Fos-related antigen protein, and is described as an anti-inflammatory and anti-oxidant compound. Phenidone has been shown to block neurotoxicity induced by kainate, and weakens oxidative stress induced by lipopolysaccharide.
Solubility
Soluble in water (1:10 hot), methanol (0.1 mg/mL), DMSO (75 mg/mL), and ethanol.
Notes
Store in cool place. Keep away from oxidizing agents.
Specifications
Melting Point | 120°C to 123°C |
Quantity | 50 g |
UN Number | UN2811 |
Beilstein | 131856 |
Merck Index | 14,7309 |
Formula Weight | 162.19 |
Percent Purity | 97% |
Chemical Name or Material | 1-Phenyl-3-pyrazolidinone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'