missing translation for 'onlineSavingsMsg'
Learn More

(±)-1-Phenyl-2-propyn-1-ol, 98+%, Thermo Scientific Chemicals

Catalog Number 10390844
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
4187-87-5
C9H8O
132.16
MFCD00021860
UIGLAZDLBZDVBL-UHFFFAOYSA-N
1-phenyl-2-propyn-1-ol, 1-phenylpropargyl alcohol, ethynylphenylcarbinol, phenylethynylcarbinol, 2-propyn-1-ol, 1-phenyl, benzyl alcohol, .alpha.-ethynyl, benzenemethanol, .alpha.-ethynyl, alpha-ethynylbenzyl alcohol, alpha-ethynylbenzenemethanol, alpha-phenylpropargyl alcohol
20155
1-phenylprop-2-yn-1-ol
C#CC(C1=CC=CC=C1)O
This item is not returnable. View return policy
4187-87-5
C9H8O
132.16
MFCD00021860
UIGLAZDLBZDVBL-UHFFFAOYSA-N
1-phenyl-2-propyn-1-ol, 1-phenylpropargyl alcohol, ethynylphenylcarbinol, phenylethynylcarbinol, 2-propyn-1-ol, 1-phenyl, benzyl alcohol, .alpha.-ethynyl, benzenemethanol, .alpha.-ethynyl, alpha-ethynylbenzyl alcohol, alpha-ethynylbenzenemethanol, alpha-phenylpropargyl alcohol
20155
1-phenylprop-2-yn-1-ol
C#CC(C1=CC=CC=C1)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point 22.0°C
Color Yellow
Density 1.0660g/mL
Boiling Point 231.0°C to 233.0°C
Flash Point 99°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula HC≡CCH(OH)C6H5
Refractive Index 1.5480 to 1.5510 (20°C, 589nm)
Quantity 1 g
Specific Gravity 1.066
Formula Weight 132.16
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material (±)-1-Phenyl-2-propyn-1-ol
Show More Show Less
compliance-icons
Product Identifier
  • (#1)-1-Phenyl-2-propyn-1-ol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'