missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-1-Phenyl-1-propanol, 98+%, Thermo Scientific Chemicals

Catalog Number p-7081033
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
93-54-9
C9H12O
136.194
MFCD00004564
DYUQAZSOFZSPHD-UHFFFAOYSA-N
1-phenyl-1-propanol, phenycholon, fepar, ejibil, felicur, livonal, 1-phenylpropanol, gallenperlen, phenylchol, bilergon
7147
1-phenylpropan-1-ol
CCC(C1=CC=CC=C1)O
This item is not returnable. View return policy
93-54-9
C9H12O
136.194
MFCD00004564
DYUQAZSOFZSPHD-UHFFFAOYSA-N
1-phenyl-1-propanol, phenycholon, fepar, ejibil, felicur, livonal, 1-phenylpropanol, gallenperlen, phenylchol, bilergon
7147
1-phenylpropan-1-ol
CCC(C1=CC=CC=C1)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.994
Boiling Point 217°C to 219°C
Flash Point 95°C (203°F)
Refractive Index 1.52
Quantity 25 g
Beilstein 1906759
Merck Index 14,3768
Formula Weight 136.19
Percent Purity ≥98%
Chemical Name or Material (±)-1-Phenyl-1-propanol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'