missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-butyne, 98%

Product Code. 11454150
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454150 25 g 25g
11444150 5 g 5g
11463734 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454150 Supplier Thermo Scientific Alfa Aesar Supplier No. A11332.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 622-76-4 | C10H10 | 130.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-76-4
Molecular Formula C10H10
Molecular Weight (g/mol) 130.19
MDL Number MFCD00039945
InChI Key FFFMSANAQQVUJA-UHFFFAOYSA-N
Synonym 1-phenyl-1-butyne, 1-butynylbenzene, benzene, 1-butynyl, 1-butyne, 1-phenyl, but-1-yn-1-ylbenzene, 1-but-1-ynyl benzene, 1-ethyl-2-phenylacetylene, phenylbutyne, ethylphenylacetylene, phenylethylacetylene
PubChem CID 69328
IUPAC Name but-1-ynylbenzene
SMILES CCC#CC1=CC=CC=C1

Specifications

Density 0.918
Boiling Point 201°C to 203°C
Flash Point 68°C (154°F)
Linear Formula CH3CH2C≡CC6H5
Refractive Index 1.552
Quantity 25 g
Beilstein 1903397
Formula Weight 130.19
Percent Purity 98%
Chemical Name or Material 1-Phenyl-1-butyne

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.