missing translation for 'onlineSavingsMsg'
Learn More

1-Pentyn-3-ol, 98%

Product Code. 11436646
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11436646 25 g 25g
11416646 1 g 1g
11426646 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11436646 Supplier Thermo Scientific Alfa Aesar Supplier No. B24853.14
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4187-86-4 | C5H8O | 84.118 g/mol

1-Pentyn-3-ol is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pentyn-3-ol is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents, heavy metal powders.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4187-86-4
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00004572
InChI Key LBSKEFWQPNVWTP-UHFFFAOYSA-N
Synonym 1-pentyn-3-ol, ethylethynylcarbinol, ethyl ethynyl carbinol, 1-pentyne-3-ol, 1-pentine-3-ol, 3-hydroxy-1-pentine, ethynyl-ethyl carbinol, acmc-1agpc
PubChem CID 92981
IUPAC Name pent-1-yn-3-ol
SMILES CCC(C#C)O

Specifications

Density 0.89
Boiling Point 123°C to 125°C
Flash Point 29°C (84°F)
Odor Recognizable
Refractive Index 1.434
Quantity 25 g
UN Number UN1986
Beilstein 1098409
Solubility Information Slightly soluble in water.
Formula Weight 84.12
Percent Purity 98%
Chemical Name or Material 1-Pentyn-3-ol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.