missing translation for 'onlineSavingsMsg'
Learn More

1-Pentylamine, 98%

Product Code. 11469163
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469163 500 g 500g
11449163 25 g 25g
11459163 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11469163 Supplier Thermo Scientific Alfa Aesar Supplier No. A11674.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-58-7 | C5H13N | 87.166 g/mol

1-Pentylamine is used as a solvent, as a raw material in the manufacture of a variety of other compounds, including dyes, emulsifiers, and pharmaceutical products and as a flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pentylamine is used as a solvent, as a raw material in the manufacture of a variety of other compounds, including dyes, emulsifiers, and pharmaceutical products and as a flavoring agent.

Solubility
Soluble in alcohol, ether.

Notes
Air Sensitive. Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-58-7
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.166
MDL Number MFCD00008236
InChI Key DPBLXKKOBLCELK-UHFFFAOYSA-N
Synonym amylamine, 1-aminopentane, pentylamine, 1-pentylamine, n-amylamine, 1-pentanamine, n-pentylamine, monoamylamine, norleucamine, amyl amine
PubChem CID 8060
ChEBI CHEBI:74848
IUPAC Name pentan-1-amine
SMILES CCCCCN

Specifications

Melting Point -55°C
Density 0.751
Boiling Point 103°C to 105°C
Flash Point −1°C (30°F)
Odor Amine-like
Refractive Index 1.411
Quantity 500 g
UN Number UN1106
Beilstein 505953
Sensitivity Air Sensitive
Merck Index 14,598
Solubility Information Soluble in alcohol,ether.
Formula Weight 87.17
Percent Purity 98%
Chemical Name or Material 1-Pentylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.