missing translation for 'onlineSavingsMsg'
Learn More

1-Pentyl isothiocyanate, 96%

Product Code. 11467177
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11467177 5 g 5g
11477177 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11467177 Supplier Thermo Scientific Alfa Aesar Supplier No. A14972.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 629-12-9 | C6H11NS | 129.221 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-12-9
Molecular Formula C6H11NS
Molecular Weight (g/mol) 129.221
MDL Number MFCD00014444
InChI Key SGHJUJBYMSVAJY-UHFFFAOYSA-N
Synonym pentyl isothiocyanate, n-amyl isothiocyanate, 1-pentyl isothiocyanate, amyl isothiocyanate, n-pentyl isothiocyanate, pentylisothiocyanate, unii-kl6u8nl15r, n-amylisothiocyanate, 1-isothiocyanato-pentane, pentane,1-isothiocyanato
PubChem CID 69415
IUPAC Name 1-isothiocyanatopentane
SMILES CCCCCN=C=S

Specifications

Density 0.93
Boiling Point 191°C to 193°C
Odor Pungent
Refractive Index 1.4965
Quantity 5 g
UN Number UN2922
Beilstein 1746691
Sensitivity Moisture sensitive
Formula Weight 129.23
Percent Purity 96%
Chemical Name or Material 1-Pentyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.