missing translation for 'onlineSavingsMsg'
Learn More

1-Penten-3-ol, 97%

Product Code. 10581181 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10581181 5 g Glass bottle 5g
10396910 25 g Glass bottle 25g
15148878 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10581181 Supplier Thermo Scientific Acros Supplier No. 130010050
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 616-25-1 | C5H10O | 86.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-25-1
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00004573
InChI Key VHVMXWZXFBOANQ-UHFFFAOYNA-N
Synonym 1-penten-3-ol, ethyl vinyl carbinol, vinyl ethyl carbinol, 1-pentene-3-ol, 1-ethylallyl alcohol, alpha-ethylallyl alcohol, 1-penten-3-ol e, 1-penten-3-ol, r, fema no. 3584, ethylvinylcarbinol
PubChem CID 12020
IUPAC Name pent-1-en-3-ol
SMILES CCC(O)C=C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow to Beige
Density 0.8390g/mL
Boiling Point 114.0°C to 115.0°C
Flash Point 25°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C2H5CH(OH)CH=CH2
Refractive Index 1.4220 to 1.4260
Quantity 5 g
Beilstein 01,443
Specific Gravity 0.839
Solubility Information Solubility in water: slightly soluble in water
Formula Weight 86.13
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1-Penten-3-ol, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.