missing translation for 'onlineSavingsMsg'
Learn More

1-Penten-3-ol, 97%

Product Code. 11460701
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460701 25 g 25g
11450701 5 g 5g
11470701 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460701 Supplier Thermo Scientific Alfa Aesar Supplier No. B20155.14
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 616-25-1 | C5H10O | 86.13 g/mol

1-Penten-3-ol is used as a flavor and a fragrance. It can be used for lift with cucumber, melon, berry, horseradish, strawberry, and tomato.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Penten-3-ol is used as a flavor and a fragrance. It can be used for lift with cucumber, melon, berry, horseradish, strawberry, and tomato.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-25-1
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00004573
InChI Key VHVMXWZXFBOANQ-UHFFFAOYNA-N
Synonym 1-penten-3-ol, ethyl vinyl carbinol, vinyl ethyl carbinol, 1-pentene-3-ol, 1-ethylallyl alcohol, alpha-ethylallyl alcohol, 1-penten-3-ol e, 1-penten-3-ol, r, fema no. 3584, ethylvinylcarbinol
PubChem CID 12020
IUPAC Name pent-1-en-3-ol
SMILES CCC(O)C=C

Specifications

Density 0.838
Boiling Point 114°C to 115°C
Flash Point 25°C (77°F)
Refractive Index 1.425
Quantity 25 g
UN Number UN1987
Beilstein 1719834
Solubility Information Slightly soluble in water.
Formula Weight 86.13
Percent Purity ≥98%
Chemical Name or Material 1-Penten-3-ol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.