missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Pentanol, ReagentPlus™, ≥99%, Honeywell™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
4 L
20 L
Packaging:
Aluminum Bottle
Glass Bottle
Stainless Steel Can
Unit Size:
1 L
100mL
2.5 L
20 L
4 L
500mL
Description
- ≥95% for general laboratory use.
Specifications
Specifications
| Melting Point | -78°C |
| Color | Colorless |
| Density | 0.811g/mL at 25°C |
| Boiling Point | 136°C to 138°C |
| Flash Point | 49°C (120.2°F) |
| Packaging | Aluminum Bottle |
| Linear Formula | CH3(CH2)4OH |
| Refractive Index | n20/D 1.409(lit.) |
| Quantity | 4 L |
| UN Number | UN1105 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction