missing translation for 'onlineSavingsMsg'
Learn More

1-Pentanol, ACS, 99+%, Thermo Scientific Chemicals

Catalog Number 11327378
Click to view available options
Quantity:
250 mL
1 L
4 L
71-41-0
C5H12O
88.15
MFCD00002977,MFCD00081734,MFCD01075169
AMQJEAYHLZJPGS-UHFFFAOYSA-N
1-pentanol, amyl alcohol, n-amyl alcohol, n-pentanol, pentanol, pentyl alcohol, butylcarbinol, 1-pentyl alcohol, amylol, n-butylcarbinol
6276
CHEBI:44884
pentan-1-ol
CCCCCO
71-41-0
C5H12O
88.15
MFCD00002977,MFCD00081734,MFCD01075169
AMQJEAYHLZJPGS-UHFFFAOYSA-N
1-pentanol, amyl alcohol, n-amyl alcohol, n-pentanol, pentanol, pentyl alcohol, butylcarbinol, 1-pentyl alcohol, amylol, n-butylcarbinol
6276
CHEBI:44884
pentan-1-ol
CCCCCO

Filtered through 0.2μ filters. Suitable for spectrophotometry, chromatography

1-Pentanol is used as a key starting material in the production of dichloro-acetic acid pentyl ester, 2-methyltetrahydrofuran, di-n-penyl ether (DNPE), pentyl butyrate and amyl acetate. As a solvent, it is used for coating CDs and DVDs. It has been considered to be a replacement for gasoline and diesel fuels due to its combustion properties. Further, it is used as a reagent to prepare derivatives of ferulic acid and caffeic acid, which shows anticancer properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -79°C
Color Colorless
Density 0.812 g/mL
Boiling Point 136°C to 138°C
Flash Point 49°C (120°F)
Odor Mild
Linear Formula CH3(CH2)4OH
Refractive Index 1.4103
Quantity 250 mL
UN Number UN1105
Beilstein 1730975
Merck Index 14,7118
Solubility Information Miscible with water,alcohol,ether,acetone and most organic solvents.
Formula Weight 88.15
Percent Purity ≥99%
Grade ACS
Physical Form Liquid
Chemical Name or Material 1-Pentanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'