missing translation for 'onlineSavingsMsg'
Learn More

1-Pentanethiol, 97%

Product Code. 11326896
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11326896 100 g 100g
11316896 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11326896 Supplier Thermo Scientific Alfa Aesar Supplier No. L04330.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 110-66-7 | C5H12S | 104.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-66-7
Molecular Formula C5H12S
Molecular Weight (g/mol) 104.21
MDL Number MFCD00004907
InChI Key ZRKMQKLGEQPLNS-UHFFFAOYSA-N
Synonym 1-pentanethiol, pentyl mercaptan, amyl mercaptan, n-amyl mercaptan, pentanethiol, amyl thioalcohol, pentalarm, amyl sulfhydrate, amyl hydrosulfide, pentylmercaptan
PubChem CID 8067
IUPAC Name pentane-1-thiol
SMILES CCCCCS

Specifications

Melting Point -76°C
Boiling Point 125°C to 126°C
Flash Point 18°C (64°F)
Odor Stench
Quantity 100 g
UN Number UN1111
Beilstein 1730979
Formula Weight 104.22
Percent Purity 97%
Chemical Name or Material 1-Pentanethiol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.