missing translation for 'onlineSavingsMsg'
Learn More

1-Pentanesulfonic acid sodium salt, monohydrate; 99.9%, For HPLC analysis, MP Biomedicals™

Product Code. 11486972 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11486972 5 g 5g
17891371 25 g 25g
17801381 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11486972 Supplier MP Biomedicals Supplier No. 0215181305
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Item Name 1-Pentanesulfonic Acid, Sodium Salt, Monohydrate
Content And Storage Store at Room Temperature.
Form Crystalline Powder
Quantity 5 g
Appearance White
Molecular Formula C5H11NaO3S
InChI Key ROBLTDOHDSGGDT-UHFFFAOYSA-M
SMILES [Na+].CCCCCS([O-])(=O)=O
IUPAC Name sodium pentane-1-sulfonate
Molecular Weight (g/mol) 174.19
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.