Learn More
1-Pentanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous, ACROS Organics™
Brand: Acros Organics 396650250
Code : 29
Additional Details : CAS Number : 22767-49-3 Weight : 0.07500kg
Packaging | Glass bottle |
---|---|
Quantity | 25g |
Description
For ion pair chromatographyChemical Identifiers
22767-49-3 | |
174.2 | |
sodium 1-pentanesulfonate, sodium pentane-1-sulfonate, sodium pentanesulfonate, 1-pentanesulfonic acid sodium salt, 1-pentanesulfonic acid, sodium salt, 1-pentylsodiumsulfonate, ipc-alks-5, 1-pentanesulfonate, sodium, pentanesulfonic acid na-salt, pentanesulfonic acidsodiumsalt | |
sodium;pentane-1-sulfonate |
C5H11O3SNa | |
ROBLTDOHDSGGDT-UHFFFAOYSA-M | |
23664617 | |
CCCCCS(=O)(=O)[O-].[Na+] |
Specifications
0.02 max. (C=0.5 M in water) at 260nm, 0.04 max. (C=0.5 M in water) at 230nm, 0.06 max. C=0.5 M in water at 220nm, 0.1 max. (C=0.5 M in water) at 210nm | |
Authentic | |
Glass bottle | |
White | |
25g | |
99+% | |
CH3(CH2)4SO3Na | |
Solubility in water: soluble. | |
CCCCCS(=O)(=O)[O-].[Na+] | |
174.2 | |
174.2 | |
99+%, |
1-Pentanesulfonic acid, sodium salt | |
2% (150°C) max. | |
(HPLC, gradient) Passes | |
>300.0°C | |
22767-49-3 | |
C5H11O3SNa | |
sodium 1-pentanesulfonate, sodium pentane-1-sulfonate, sodium pentanesulfonate, 1-pentanesulfonic acid sodium salt, 1-pentanesulfonic acid, sodium salt, 1-pentylsodiumsulfonate, ipc-alks-5, 1-pentanesulfonate, sodium, pentanesulfonic acid na-salt, pentanesulfonic acidsodiumsalt | |
ROBLTDOHDSGGDT-UHFFFAOYSA-M | |
sodium;pentane-1-sulfonate | |
23664617 | |
Crystalline Powder | |
Ion Pair Chromatography |
Safety and Handling
- 1-Pentanesulfonic acid, sodium salt
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.