missing translation for 'onlineSavingsMsg'
Learn More

1-Pentadecene, 97%

Product Code. 15420428
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15420428 25 g 25g
15410428 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15420428 Supplier Thermo Scientific Alfa Aesar Supplier No. H53516.14
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13360-61-7 | C15H30 | 210.405 g/mol

1-Pentadecene serves as a growth medium for the strain CV2803T. It is a common semiochemical of flour beetles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pentadecene serves as a growth medium for the strain CV2803T. It is a common semiochemical of flour beetles.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13360-61-7
Molecular Formula C15H30
Molecular Weight (g/mol) 210.405
MDL Number MFCD00008987
InChI Key PJLHTVIBELQURV-UHFFFAOYSA-N
Synonym 1-pentadecene, pentadecene, pentadecene,1, unii-kh577lia74, ccris 5721, acmc-1bt6q
PubChem CID 25913
ChEBI CHEBI:77506
IUPAC Name pentadec-1-ene
SMILES CCCCCCCCCCCCCC=C

Specifications

Melting Point -4°C
Density 0.775
Boiling Point 268°C to 269°C
Flash Point 110°C (230°F)
Linear Formula CH3(CH2)12CH=CH2
Refractive Index 1.439
Quantity 25 g
Beilstein 1751011
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 210.4
Percent Purity 97%
Chemical Name or Material 1-Pentadecene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.