Learn More
1-Pentadecene, 97%
Description
1-Pentadecene serves as a growth medium for the strain CV2803T. It is a common semiochemical of flour beetles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Pentadecene serves as a growth medium for the strain CV2803T. It is a common semiochemical of flour beetles.
Solubility
Not miscible or difficult to mix in water.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | -4°C |
| Density | 0.775 |
| Boiling Point | 268°C to 269°C |
| Flash Point | 110°C (230°F) |
| Linear Formula | CH3(CH2)12CH=CH2 |
| Refractive Index | 1.439 |
| Quantity | 5 g |
| Beilstein | 1751011 |
| Solubility Information | Not miscible or difficult to mix in water. |
| Formula Weight | 210.4 |
| Show More |
RUO – Research Use Only
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.