missing translation for 'onlineSavingsMsg'
Learn More

1-Palmitoyl-sn-glycero-3-phosphocholine, TRC

Product Code. 17998684
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17998684 50 mg 50mg
17917934 100 mg 100mg
17997454 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17998684 Supplier Toronto Research Chemicals Supplier No. TRCP15734550MG
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17364-16-8
Molecular Formula C24H50NO7P
Molecular Weight (g/mol) 495.63
Synonym (7R)-3,5,9-4,7-Dihydroxy-N,N,N-trimethyl-10-oxotrioxa-4-phosphapentacosan-1-aminium 4-Oxide Inner Salt, 1-Hexadecanoyl-sn-glycerol-3-phosphorylcholine, 1-Hexadecanoyllysolecithin, 1-O-Palmitoyl-2-lyso-sn-glycero-3-phosphocholine, MPPC, Palmitoyl L-Lysophosphatidylcholine, Palmitoyl L-α-Lysolecithin, Palmitoyl L-α-Lysophosphatidylcholine, γ-Palmitoyl-L-α-lysophosphatidylcholine;
SMILES O=P([O-])(OCC[N+](C)(C)C)OC[C@H](O)COC(CCCCCCCCCCCCCCC)=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C24H50NO7P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-24(27)30-21-23(26)22-32-33(28,29)31-20-19-25(2,3)4/h23,26H,5-22H2,1-4H3/t23-/m1/s1
Formula Weight 495.3325
Chemical Name or Material 1-Palmitoyl-sn-glycero-3-phosphocholine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.