missing translation for 'onlineSavingsMsg'
Learn More

1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine (main component of Egg Phosphatidylcholines / EPC, CAS 97281-44-2), Mikromol™

Product Code. 17967214
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17967214 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17967214 Supplier Mikromol Supplier No. MM3893.030025
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine (main component of Egg Phosphatidylcholines / EPC, CAS 97281-44-2)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Palmitoyl-2-oleoyl-sn-glycerol-3-phosphocholine
CAS 26853-31-6
Analyte or Component Names 1-Palmitoyl-2-oleoyl-sn-glycerol-3-phosphocholine
Shipping Condition Ice pack (-20°C)
Molecular Formula C42H82NO8P
Certifications/Compliance ISO 17025
SMILES CCCCCCCCCCCCCCCC(=O)OC[C@H](COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCC\C=C/CCCCCCCC
Molecular Weight (g/mol) 760.08
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.